Learn More
Digital Laser Power Meter

Shop All Industrial Fiber Optics Inc Products


Unit features settings for standard and peak/hold radiometric measurements and percentage mode for calibrating/measuring to user-defined standards.
- Easy-to-read digital readout
- Zero offset adjustment to compensate for ambient light
- With radiometric mode in 100 percent offset control; displays readings directly as percentages
- Detachable detector head with threaded mounting hole for use on a ringstand or optical bench
- Calibration charts and instruction guides included
- Dual-powered (110 VAC or two 9V batteries for portability)
Specifications
Product Type
Digital Laser Power Meter
Description
LCD display; Operating Temperature: 10° to 30°C (50° to 86°F)
Electrical Requirements
110 VAC 2 w
For Use With (Application)
Research applications and cdrh compliance measurements
Weight (English)
2.2 lb.
Weight (Metric)
1 kg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.