Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inorganic Carbon Standard, 50 ppm C, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
Color | Colorless |
Assay Percent Range | 50 ± 1ppm C |
Odor | Odorless |
Solvent or Matrix | Aqueous Solution |
Solubility | Infinitely soluble in water |
Physical Form | Liquid |
Chemical Name or Material | Inorganic Carbon Standard, 1mL = 0.5mg C, 50ppm C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction