Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Inorganic Carbon Standard, 1000 ppm C, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
Packaging | Amber Glass Bottle |
Quantity | 120 mL |
CAS Min % | 97.0, 0.43, 0.34 |
CAS Max % | 100.0, 0.45, 0.35 |
Color | Colorless |
Concentration or Composition Notes | 1000ppm C |
Equivalence | 1 mL = 1 mg C |
Physical Form | Liquid |
Chemical Name or Material | Inorganic Carbon Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction