Learn More
BioMed Diagnostics InPouch TV is a self-contained broth mediadevice for the recovery and detection ofTrichnomonas vaginalis from female vaginalsamples or male urethra/urine samples. Available in: 10 pack
Supplier:  BioMed Diagnostics 11031001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
InPouch TV is a self-contained broth media device for the recovery and detection of Trichnomonas vaginalis from female vaginal samples or male urethra/urine samples. The selective, nutritive media provides presumptive-positive results of one or more live trichomonads under the microscope immediately after inoculation of specimen. Suitable for use with urine, vaginal swab, and seminal fluid. | Detects: Trichnomonas vaginalis from female vaginal samples or male urethra/urine samples. | Available in: 10 pack
Catalog No. NC2131084