Learn More
Kamiya Biomedical Insulin Reagent (Large Size), 27mL+10mL/PK RX
Supplier: Kamiya Biomedical KAI071
K-ASSAY Insulin (L) Assay Reagent. R1: 2 x 13.5mL (liquid), R2: 2 x 5mL (liquid). Assay Range: 1 - 100 IU/mL. For the quantitative determination of human insulin in serum and plasma by immunoturbidimetric assay. FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY Insulin Calibrator (Cat. No. KAI-072C). Recommended Controls: K-ASSAY Insulin Control (Cat. No. K73C-2M or K73C-4M). This assay can be used on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.