Learn More
Honeywell IntelliDoX Docking Station
Honeywell IntelliDoX Docking Station is a bump test and calibration station for gas detectors that demonstrates quick and efficient performance.
$4790.00 - $4800.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
19-807-579
|
Honeywell
DXMAXXT |
BW MAXXT |
Each for $4,790.00
|
|
|||||
19-807-578
|
Honeywell
DXBWC4 |
Each for $4,800.00
|
|
||||||
19-807-577
|
Honeywell
DXMCLP |
BW MicroClip Series |
Each for $4,800.00
|
|
|||||
19-807-575
|
Honeywell
DXSOLO |
BW Solo |
Each for $4,790.00
|
|
|||||
Description
Honeywell IntelliDoX Docking Station is a bump test and calibration station for gas detectors that demonstrates quick and efficient performance.
- High-tech way to combine smart docking modules and Fleet Manager II software to provide automated instrument management to drive productivity, reliability and efficiency.
- Enables bump test tracking, email notices and easy recordkeeping for improved compliance
- Performs a quick bump test for increased uptime and no wasted calibration gas
- Enables the ability to change alarm set points, set a noncompliance indicator when a bump test is due, and adjust other parameters for maximum flexibility
- IntelliDoX module comes with factory-installed nest
- Power cord, calibration gas tubing and exhaust tubing are packaged separately
- One Enabler Kit is required for each stand-alone IntelliDoX module
- 2-year warranty