Learn More
Inorganic Ventures 200.7 82 INTERFER CHK 3 125mL (Inorganic Ventures)
SDP
Supplier:  Inorganic Ventures 2007ICS3125ML
Encompass_Preferred
Matrix: Nitric Acid (HNO3), Dilution: 1:100, Analyte: As 1000 ug/mL, Be 100 ug/mL, Cr & Cu 300 ug/mL, Mn 200 ug/mL, K 20000 ug/mL, Ag, V, Ba, Cd, Co, Ni, Zn all at 300 ug/mL, Pb 1000 ug/mL, Se 500 ug/mL, and Tl 1000 ug/mL.
Catalog No. 50-723-001
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.