Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Chemicals Interleukin 2, human
$175.20
Specifications
Molecular Weight (g/mol) | 15.5 kd |
---|---|
Packaging | Plastic bottle |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ67298EXE
|
Thermo Scientific Chemicals
J67298EXE |
10μg |
Each for $175.20
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ67298IAM
|
Thermo Scientific Chemicals
J67298IAM |
N/A | N/A | N/A | |||||
AAJ67298MCR
|
Thermo Scientific Chemicals
J67298MCR |
1mg | N/A | N/A | N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
15.5 kd | |
Plastic bottle | |
Lyophilized |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title