Learn More
MilliporeSigma™ Supelco™ Intermediate Polar Fused Silica Tubing
Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene
Supplier:  MilliporeSigma™ Supelco™ 25787
 View more versions of this product
Catalog No. 11-100-3597
missing translation for 'QuantityLeft'
Add to Cart
Description
Description
  • Use as transfer lines, guard columns, or retention gaps
  • Can be used to make custom columns
  • Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene
  • Composition: Phenyl/Methyl
  • Deactivated
Specifications
Specifications
Injection Solvents Such as Acetone, Methylene Chloride (Dichloromethane), and Toluene
Use as Transfer Lines, Guard Columns, Retention Gaps, or Custom Columns
60 m
Tubing
0.25 mm
Silica
Product Suggestions
Product Suggestions