Learn More
MilliporeSigma™ Supelco™ Intermediate Polar Fused Silica Tubing
Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene
Supplier:  MilliporeSigma™ Supelco™ 25745U
 View more versions of this product
Catalog No. 11-100-4715
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • Use as transfer lines, guard columns, or retention gaps
  • Can be used to make custom columns
  • Ideal for use with injection solvents such as acetone, methylene chloride (dichloromethane), and toluene
  • Composition: Phenyl/Methyl
  • Deactivated
Specifications
Specifications
Injection Solvents Such as Acetone, Methylene Chloride (Dichloromethane), and Toluene
Use as Transfer Lines, Guard Columns, Retention Gaps, or Custom Columns
5 m
Tubing
0.10 mm
Silica
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.