Learn More
Invitrogen™ pCR™8/GW/TOPO™ TA Cloning Kit with One Shot™ TOP10 Chemically Competent E. coli (Supply Center Only)

Includes
TOPO-adapted, ready-to-use pCR8/GW/TOPO Vector, dNTPs, Salt Solution, Sterile Water, GW Forward and Reverse Sequencing Primers, Control Template and PCR Primers, TOP10 Chemically Competent E. coli or Mach1-T1R Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. The PureLink Quick Plasmid Miniprep Kit contains Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffer, TE Buffer, Spin Columns, and Wash and Recovery Tubes.
- Use pCR™-TOPO Vector with covalently bound topoisomerase I for fast cloning and recombinants
- Comes with Gateway (GW) entry vector, pCR™8/GW/TOPO
- Vector has 3'-T overhangs for direct ligation of Taq-amplified PCR products
- att sites for rapid recombination into variety of Gateway destination vectors
- Novel GW primer sites, within the att regions, located less than 55 base pairs from the PCR product insertion site for convenient sequencing
- Spectinomycin resistance gene for robust selection in E. coli
- EcoR I sites flanking PCR product insertion site for easy excision of inserts
Cloning,Gateway Cloning,PCR Cloning
Order Info
Shipping Condition: Dry Ice
Specifications
Cloning Method
TOPO™ with Gateway™
Content And Storage
The pCR™8/GW/TOPO™ TA Cloning Kit contains the TOPO™-adapted, ready-to-use pCR™8/GW/TOPO™ Vector, dNTPs, Salt Solution, Sterile Water, GW Forward and Reverse Sequencing Primers, Control Template and PCR Primers, One Shot™ TOP10 Chemically Competent E. coli or One Shot™ Mach1™-T1R Chemically Competent E. coli, S.O.C. Medium, and a pUC19 control plasmid. The PureLink™ Quick Plasmid Miniprep Kit contains Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffer, TE Buffer, Spin Columns, and Wash and Recovery Tubes. Store all components of the PureLink™ kit at room temperature. Store the One Shot™ E. coli at -80°C. Store all other components at -20°C. All reagents are guaranteed stable for 6 months when properly stored.
No. of Reactions
20 reactions
Product Line
One Shot™
Product Type
TA Cloning Kit
Shipping Condition
Dry Ice
Vector
pCR
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.