Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Terminal Deoxynucleotidyl Transferase, recombinant
SDP

Performance and quality tested

$291.65 - $622.65

Specifications

Content And Storage Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C.
Enzyme Terminal Deoxynucleotidyl Transferase
Compatible Buffer 5X Buffer, Reaction Buffer
Product Type TdT
View More Specs

Alert:

Hazard Warning: Toxic; potassium cacodylate contained in reaction buffer. Also contains cobalt chloride, a highly toxic chemical. See MSDS.


Includes: Vial of 5X buffer [500mM potassium cacodylate (pH 7.2), 10mM CoCl2 , 1mM DTT]

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
10-533-065
SDP
Encompass
View Documents
Invitrogen™
10533065
500 U
Each for $291.65

Save 7%

Reg : $312.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
 
10-533-073
SDP
Encompass
View Documents
Invitrogen™
10533073
3 x 500 U
Each for $622.65

Save 7%

Reg : $666.00

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
 
Description

Description

Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is DNA polymerase that catalyzes addition of deoxynucleotides to 3fi hydroxyl terminus of DNA.

  • Source: Purified from baculovirus clone of calf thymus TdT
  • Performance and Quality Testing: Endonuclease, 3' and 5' exodeoxyribonuclease, and levels of incorporation tested
  • Unit Definition: One unit incorporates 1nmol dATP into acid-precipitable material in 1 hr. at 37°C, using d(pA)50 as primer
  • Unit Reaction Conditions: 0.2M potassium cacodylate (pH 7.2), 10mM MgO4 C4 H6 , 1mM 2-mercaptoethanol, 0.5mg/mL BSA, 100 flM d(pA)50, 1mM [3H]dATP, and enzyme in 0.15mL for 1 h at 37°C

Cloning; Restriction Enzyme Cloning; Homopolymer tailing of vector and insert for cloning; Labeling oligonucleotides with biotin, 32P- or 35S-label; Apoptosis (TUNEL)

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

Specifications

Specifications

Terminal Deoxynucleotidyl Transferase, Recombinant (rTdT) is supplied with vial of 5X buffer [500 mM potassium cacodylate (pH 7.2), 10 mM CoCl2 , 1 mM DTT]. Store at -20°C.
Terminal Deoxynucleotidyl Transferase
5X Buffer, Reaction Buffer
TdT
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.