Learn More
Bellco Glass, Inc. Iob W/ Micro-Carrier Flask 1L
Supplier: Bellco Glass, Inc. 196561010
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. Cap size: 100mm Flask size: 1l Dimensions: 135 x 250mm Sidearm cap: 45mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.