Learn More
Bellco Glass, Inc. Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. Cap size: 100mm Flask size: 500ml Dimensions: 110 x 180mm Sidearm cap: 45mm
SDP
Supplier:  Bellco Glass, Inc. 196561015
Encompass_Preferred
Internal impeller decreases risk of contamination. Impeller has an adjustable shaft to align paddle. The large paddle promotes increased oxygenation. Cap size: 100mm Flask size: 500ml Dimensions: 110 x 180mm Sidearm cap: 45mm
Catalog No. 50-194-5061
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.