Learn More
Bellco Glass, Inc. Internal impeller is useful because it decreases risk of contamination. Has an adjustable shaft to align the paddle. Large paddle promotes increased oxygenation. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. The seal
SDP
Supplier:  Bellco Glass, Inc. 196562001
Encompass_Preferred
Internal impeller is useful because it decreases risk of contamination. Has an adjustable shaft to align the paddle. Large paddle promotes increased oxygenation. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. The seal
Catalog No. 50-194-5067
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.