Learn More
Bellco Glass, Inc. Internal impeller is useful because it decreases risk of contamination. Has an adjustable shaft to align the paddle. Large paddle promotes increased oxygenation. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. The seal
Supplier: Bellco Glass, Inc. 196562005
Internal impeller is useful because it decreases risk of contamination. Has an adjustable shaft to align the paddle. Large paddle promotes increased oxygenation. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. The seal
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.