Learn More
Bellco Glass, Inc. Internal impeller is useful because it decreases risk of contamination. Has an adjustable shaft to align the paddle. Large paddle promotes increased oxygenation. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. The seal
Internal impeller is useful because it decreases risk of contamination. Has an adjustable shaft to align the paddle. Large paddle promotes increased oxygenation. Stainless steel shaft design prevents cell damage and minimizes magnetic decoupling. The seal