Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, 0.01 Normal (N/100) Volumetric Solution, StandARd™, Macron Fine Chemicals™

Catalog No. MKH1751 Shop All Avantor Products
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
PubChem CID 807
ChEBI CHEBI:17606
IUPAC Name diiodine
SMILES II

Specifications

Packaging Narrow Mouth Amber Glass Bottle
Quantity 1 L
Grade StandARd™
Specific Gravity 1L = 1.00kg
Chemical Name or Material Iodine, 0.01 Normal (N/100) Volumetric Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.