Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine-125, MP Biomedical

Catalog No. MP016303725 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

10034-85-2

Molecular Formula

HI

Molecular Weight (g/mol)

127.91

MDL Number

MFCD00011347

InChI Key

XMBWDFGMSWQBCA-UHFFFAOYSA-N

Synonym

hydriodic acid, hydrogen iodide, hydroiodic acid, hydriotic acid, iodium, yodo, jodwasserstoff, iodine-125, unii-694c0eft9q, iodo group

PubChem CID

24841

ChEBI

CHEBI:43451

IUPAC Name

hydrogen iodide

SMILES

I

Specifications

Quantity

25 mci

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.