Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071847 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
500g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent, solid,≥99.8%

Honeywell ACS solvents for general laboratory use are:

  • a value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested tmeet ACS general use specifications

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Physical Form Solid
Quantity 100 g
UN Number 3495
Beilstein 3587194
IUPAC Name diiodine
Formula Weight 253.81g/mol
Percent Purity ≥99.8%
Grade ACS Reagent
Vapor Pressure 0.31mmHg (25°C), 1mmHg (38.7°C)
Density Vapor Density: 9 (vs air)
Chemical Name or Material Iodine
Show More Show Less
Hazard Statement P261-P273-P280-P305 + P351 + P338-P314
Precautionary Statement H312 + H332-H315-H319-H335-H372-H400
EINECSNumber 231-442-4
RTECSNumber NN1575000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.