Learn More
Volu Sol Iodine Alcohol (Ethanol 70% Dilutant Included), 80 mL & 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VXI128
Encompass_Preferred
Iodine Alcohol, commonly used to remove the mercury deposits in Schaudinn or PVA fixed fecal smears.  Prevents mercury-caused artifacts on slides during the Wheatley Trichrome staining procedure.  Component of Volu-Sol's Parasitology Kit.  This is a two-part item
Catalog No. 50-420-830
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.