Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine (Iodine-Iodide), 0.00315 N, Ricca Chemical
SDP

Catalog No. R39703004C Shop All Ricca Chemical Company Products
Click to view available options
:
Package: Amber Glass; Size: 4 L

Specifications

Chemical Name or Material Iodine
Boiling Point 100°C
Melting Point 0°C
Quantity 4 L
CAS 7553-56-2, 7681-11-0, 7732-18-5
CAS Min % 0.04, 0.13, 99.83
CAS Max % 0.04, 0.13, 99.83
Assay Percent Range 0.0031 to 0.0032
Assay Percent Notes Normality Iodine
Packaging Amber Glass Bottle
Appearance Dark Brown Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.