Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine Monochloride Solution, Wijs, for Iodine Absorption Number of Fats and Oils, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Safety-coated Glass Bottle
Specifications
Specifications
CAS Min % | 96.0, 1.41, 0.09 |
CAS Max % | 100.0, 1.46, 0.1 |
Packaging | Amber Glass Bottle |
Quantity | 100 mL |
Chemical Name or Material | Iodine Monochloride Solution, Wijs |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction