Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Potassium Iodide, 1.8%, Dilute Lugol's
SCI_ED

Catalog No. p-4666453 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
1.80%

Chemical Identifiers

CAS 7681-11-0, 7553-56-2, 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Brown
Physical Form Liquid
Quantity 100 mL
Linear Formula I2/KI
Grade Potassium Iodide, 2% w/v; Iodine, 18% w/v; Water, purified, >96% w/v
Packaging 1.80%
Density 1 to 1.05
Chemical Name or Material Iodine Potassium Iodide, 1.8%, Dilute Lugol's
Recommended Storage Stable under normal conditions of use and storage
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.