Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine, Resublimed p.a., Thermo Scientific Chemicals

Catalog No. p-4509780
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
PubChem CID 807
ChEBI CHEBI:17606
SMILES II

Specifications

Boiling Point 185°C
Melting Point 113°C
Color Gray
Physical Form Flakes or Spheres
Quantity 100 g
Assay Percent Range 99.5%
Linear Formula I2
Fieser 01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153
Merck Index 15, 5057
IUPAC Name diiodine
Formula Weight 253.81
Percent Purity ≥99.4% (Iodometry)
Grade Extra Pure
Packaging Glass Bottle
Vapor Pressure 0.41hPa at 25°C
Density 8.8 (Air = 1.0)
Chemical Name or Material Iodine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove to fresh and keep confortable for several minutes.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.