Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodobenzene 99.0+%, TCI America™
SDP

Catalog No. p-7131179 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6H5I
Molecular Weight (g/mol) 204.01
MDL Number MFCD00001029
InChI Key SNHMUERNLJLMHN-UHFFFAOYSA-N
Synonym benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene
PubChem CID 11575
IUPAC Name iodobenzene
SMILES IC1=CC=CC=C1

Specifications

Melting Point -29°C
Color Yellow
Boiling Point 188°C
Quantity 25 g
Formula Weight 204.01
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Iodobenzene
EINECSNumber (9)-1501
RTECSNumber DA3390000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.