Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodobromide TS, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
Chemical Name or Material | Iodobromide |
Color | Orange-Red |
Name Note | Test Solution |
CAS | 7789-33-5, 64-19-7 |
CAS Min % | 96.0, 1.83 |
CAS Max % | 100.0, 1.91 |
Molecular Formula | BrI |
MDL Number | MFCD00011353 |
Synonym | iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution |
InChI Key | CBEQRNSPHCCXSH-UHFFFAOYSA-N |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction