Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ion Chromatography Fluoride Standard, 100μg/mL F-, SPEX CertiPrep™
Shop All Spex CertiPrep Products

Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
Triple-leached LDPE bottle
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
F&superscript_minus; | 100μg/mL |
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Packaging | Triple-leached LDPE bottle |
Quantity | 125 mL |
CAS Min % | 0.01, 99.99 |
CAS Max % | 0.01, 99.99 |
Certificate of Analysis | With Certificate of Analysis. |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Concentration or Composition (by Analyte or Components) | 100μg/mL F− |
Solvent or Matrix | H2O |
Chemical Name or Material | Fluoride Standard |
Safety and Handling
DOTInformation | Nonhazardous |
Recommended Storage | Ambient |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction