Learn More
Phosphate-P Standard for Ion Chromatography, SPEX CertiPrep™

Shop All Spex CertiPrep Products


Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
PO&subscript_4;-P | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Packaging | Triple-leached LDPE bottle |
Quantity | 125 mL |
CAS Min % | 0.1, 99.9 |
CAS Max % | 0.1, 99.9 |
CAS | 7732-18-5 |
Certificate of Analysis | With Certificate of Analysis |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Concentration or Composition (by Analyte or Components) | 10002g/mL |
Solvent or Matrix | H2O |
Show More |
Safety and Handling
DOTInformation | Nonhazardous |
Recommended Storage | Ambient |
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.