Learn More
Biocare Medical, LLC Ion-Exchange Decal (IED) 500ML
SDP
Supplier:  Biocare Medical, LLC IED1204
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Ion-Exchange Decal Unit removes calcium from bone quickly while leaving superior cellular detail. It incorporates a strong cation ion-exchange resin in a weak acid solution to replace calcium ions with hydrogen ions.
Catalog No. 50-828-91