Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IOX 1, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
IOX 1 is a histone demethylase JMJD inhibitor (IC50 values are 0.12, 0.17, 0.2, 0.3, 0.6 and 1 μM for JMJD3, JMJD1A, JMJD2A, JMJD2E, JMJD2C and UTX respectively). Cell permeable; it inhibits JMJD2A-mediated H3K9me3 demethylation in HeLa cells. IOX 1 suppresses IL-17 expression in mouse and human CD4+ T cells by targeting TET2. It also reduces infiltration of Th17 cells in an in vivo model of intraocular inflammation.
Specifications
Specifications
Quantity | 10 mg |
Synonym | iox1, 8-hydroxy-5-quinolinecarboxylic acid, 5-carboxy-8-hydroxyquinoline, iox 1, unii-jm015yqc1c, iox-1, 5-carboxy-8hq, 5-quinolinecarboxylic acid, 8-hydroxy, jm015yqc1c, 4bio |
Formula Weight | 189.17 |
Percent Purity | >99% |
Chemical Name or Material | IOX 1 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction