Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contec™ IPA
Contec™ Isopropanol contains USP grade isopropanol (IPA) and USP grade purified water.
Supplier: Contec™ SB327030
This item is not returnable.
View return policy
Description
- Filtered to 0.2 μm, bottles are double bagged to assure product quality.
- Each bottle has a lot number code and expiration date for easy record keeping
- Evaporates and leaves extremely low residues
- Filtered through 0.2 micron in-line filter eliminates the need for in-house filtration
- Meets USP grade purified water
- Convenient trigger sprayer with settings for stream or mist spraying is included with the 16 and 32 oz. bottles - Stream setting is ideal for targeting specific hard to reach areas, while the mist setting is used to cover larger surfaces.
- Each leak-proof bottle is induction sealed, capped and shipped with a trigger sprayer to eliminate the occurrence of leakage.
- Product applications include surface cleaning, wiping gloved hands in sterile suites or during gowning, and wiping down products and equipment for pass through to controlled environments.
Specifications
Non-sterile | |
70% UPS grade Isopropyl Alcohol | |
12 Bottles/Cs. | |
0.95 L |
Clear | |
Plastic Bottle | |
32 oz. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction