Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gold Biotechnology Inc IPTG | 367-93-1 | MFCD00063273 | 5 g
SDP

Supplier:  Gold Biotechnology Inc I2481C5

Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred

IPTG | Purity: 99% | Mol Wt: 238.30 | 367-93-1 | MFCD00063273 | 5 g

Catalog No. 50-153-2804


missing translation for 'QuantityLeft'
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.