Learn More
Agilent Technologies IPTG
Supplier:  Agilent Technologies 300127
Encompass
IPTG is used in combination with X-gal to detect beta-galactosidase activity with a white/blue color assay and also for screening of transformed colonies.
Catalog No. 50-125-518
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.