Learn More
Gold Biotechnology Inc IPTG EZ Pak™ for 1 M Solution 10 mL
SDP
Supplier:  Gold Biotechnology Inc I2481EZ10
Encompass
IPTG is an analog of galactose that is non-metabolizable and inactivates the lac repressor to induce synthesis of β-galactosidase in E. coli. The expression of cloned genes under the control of the lac operon is induced by IPTG. It is also a substrate for thigalactoside transacetylase and has been reported to induce penicillinase in bacteria.
Catalog No. NC2596278
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.