Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tingley™ Iron Eagle™ Polyurethane Rainwear: Bib Overalls

Catalog No. p-9056034 Shop All Tingley Rubber Co. Products
Click to view available options
Size:
Small
Medium
This item is not returnable. View return policy
This item is not returnable. View return policy

Extremely comfortable and flexible.

  • Lightweight and durable. Resists fats, oils, pine tars, gasoline, grease and a variety of other chemicals.
  • Snag and abrasion resistant
  • Heat sealed seams
  • Therm-O-Rad™ sealed seams
  • Storm fly front
  • Premium snap-lock suspender buckles
  • Heavy-duty suspenders with crossover
  • Heavy-duty snaps
  • Mildew resistant

Order Info

Overalls available in sizes Small to 6X and jacket in sizes Small to 4X. Contact your Fisher Safety Customer Service Representative for ordering information on sizes not shown.

Specifications

Size Medium
Suspender Snap-lock
Length (English) Inseam 29 in.
Length (Metric) Inseam 73.6 cm
Size (English) Waist 36 to 38 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.