Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) Phthalocyanine (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. p-7131595 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 132-16-1
Molecular Formula C32H16FeN8
Molecular Weight (g/mol) 568.38
MDL Number MFCD00015953
InChI Key MIINHRNQLVVCEW-UHFFFAOYSA-N
Synonym iron phthalocyanine, iron ii phthalocyanine, ferrous phthalocyanine, phthalocyanine iron ii, fepc, 29h,31h-phthalocyaninato 2--n29,n30,n31,n32 iron, iron, 29h,31h-phthalocyaninato 2--n29,n39,n31,n32-, sp-4-1, iron, 29h,31h-phthalocyaninato 2--kappan29,kappan30,kappan31,kappan32-, sp-4-1
PubChem CID 2735065
IUPAC Name λ²-iron(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19,21(38),22,24,26,28,30,32,34-nonadecaene-37,39-diide
SMILES [Fe++].[N-]1C2=NC3=NC(=NC4=C5C=CC=CC5=C([N-]4)N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31

Specifications

Color Purple
Quantity 200 mg
Formula Weight 568.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iron(II) Phthalocyanine (purified by sublimation)
EINECSNumber (5)-4130
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.