Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Iron Metal Powder
SCI_ED

Catalog No. p-4416390 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Powder
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Black to Gray
Density 7.86g/mL at 20°C
Assay Percent Range >97% w/v
Linear Formula Fe
Quantity 100 g
Formula Weight 55.85
Grade Laboratory
Packaging Powder
Chemical Name or Material Iron Metal Powder
ShelfLife 5 years
DOTInformation DOT Class 4.1, II, : Flammable Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.