Learn More
Iron powder, 98% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Gray |
Appearance | Gray powder |
Density | 7.87g/cm3 at 20°C |
Assay Percent Range | 98% (Metals basis) |
MDL Number | MFCD00010999 |
Quantity | 250 g |
Mesh Size | -325 Mesh |
Form | Powder |
Chemical Name or Material | Iron, Reduced |
Safety and Handling
Hazard Category | H500-X |
Precautionary Statement | P235+P410-P280 |
DOTInformation | DOT Class: 4.2; Packaging Group: II |
EINECSNumber | 231-096-4 |
RTECSNumber | NO4565500 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.