Learn More
TARGETMOL CHEMICALS INC ISAM-140 5MG
Supplier:  TARGETMOL CHEMICALS INC T27629
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ISAM-140 is a potent and highly selective antagonist of A2B adenosine receptor. purity: 97%
Catalog No. 50-258-0594
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.