Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoamyl 2-Furancarboxylate (contains 2-Methylbutyl 2-Furancarboxylate) 80.0+%, TCI America™
SDP

Catalog No. p-7129510 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 615-12-3
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00059015
InChI Key FPBZWZKAUPNMMV-UHFFFAOYSA-N
Synonym 2-Furancarboxylic Acid Isoamyl Ester, Isoamyl 2-Furoate, 2-Furoic Acid Isoamyl Ester
PubChem CID 69203
IUPAC Name 3-methylbutyl furan-2-carboxylate
SMILES CC(C)CCOC(=O)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 232°C
Quantity 25 mL
Formula Weight 182.22
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Isoamyl 2-Furancarboxylate (contains 2-Methylbutyl 2-Furancarboxylate)
EINECSNumber (5)-0046
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.