Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog No. p-4228751
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35432-36-1
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.63
InChI Key NQLNHTOCFWUYQE-UHFFFAOYSA-N
Synonym isobutanesulfonyl chloride, isobutylsulfonyl chloride, 1-propanesulfonyl chloride, 2-methyl, 2-methylpropane-1-sulfonylchloride, 2-methyl-1-propylsulfonyl chloride, 2-methyl-1-propanesulfonyl chloride, 2-methyl-propane-1-sulfonyl chloride, isobutane sulfonyl chloride, isobutane-1-sulfonylchloride, acmc-1ae06
PubChem CID 182236
IUPAC Name 2-methylpropane-1-sulfonyl chloride
SMILES CC(C)CS(=O)(=O)Cl

Specifications

Color Brown to Colorless
Boiling Point 77°C to 78°C (11.0 Torr)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 156.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Isobutanesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.