Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isobutyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. I009825ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 592-65-4
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.292
MDL Number MFCD00039852
InChI Key CMWSRWTXVQLHNX-UHFFFAOYSA-N
Synonym Diisobutyl Sulfide
PubChem CID 11608
IUPAC Name 2-methyl-1-(2-methylpropylsulfanyl)propane
SMILES CC(C)CSCC(C)C

Specifications

Color Colorless
Boiling Point 170°C
Quantity 25 mL
UN Number 1993
Formula Weight 146.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isobutyl Sulfide
EINECSNumber (2)-0474
RTECSNumber WQ4900000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.