Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyl Cinnamate 98.0+%, TCI America™
SDP

Catalog No. C177925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 7780-06-5
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.24
MDL Number MFCD00048292
InChI Key RGACABDFLVLVCT-CMDGGOBGSA-N
Synonym isopropyl cinnamate, cinnamic acid, isopropyl ester, iso-propyl cinnamate, 2-propenoic acid, 3-phenyl-, 1-methylethyl ester, 1-methylethyl 3-phenylpropenoate, unii-6rwc10bdd7, isopropyl 3-phenylpropenoate, fema no. 2939, trans-cinnamic acid isopropyl ester, isopropyl 3-phenylacrylate
PubChem CID 5273464
IUPAC Name propan-2-yl (E)-3-phenylprop-2-enoate
SMILES CC(C)OC(=O)C=CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 270°C
Quantity 25 g
Formula Weight 190.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Cinnamate
EINECSNumber (3)-1750
RTECSNumber GD9625000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.