Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyldiphenylphosphine 97.0+%, TCI America™
SDP

Catalog No. p-7131499 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 6372-40-3
Molecular Formula C15H17P
Molecular Weight (g/mol) 228.28
MDL Number MFCD00015028
InChI Key LLZAIAIZAVMQIG-UHFFFAOYSA-N
Synonym isopropyldiphenylphosphine, phosphine, 1-methylethyl diphenyl, isopropyldiphenylphosphane, isopropyl diphenyl phosphine, acmc-209nho, isopropyl-diphenylphosphine, diphenyl propan-2-yl phosphane, i-propyldiphenylphosphine
PubChem CID 80754
IUPAC Name diphenyl(propan-2-yl)phosphane
SMILES CC(C)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 40°C
Color White
Quantity 1 g
Formula Weight 228.27
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropyldiphenylphosphine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.