Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropyltriphenylphosphonium Iodide 98.0+%, TCI America™
SDP

Catalog No. p-7131479 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 24470-78-8
Molecular Formula C21H22IP
Molecular Weight (g/mol) 432.29
MDL Number MFCD00031595
InChI Key HHBXWXJLQYJJBW-UHFFFAOYSA-M
Synonym isopropyltriphenylphosphonium iodide, phosphonium, 1-methylethyl triphenyl-, iodide, isopropyltriphenylphosphanium iodide, isopropyl triphenyl phosphonium iodide, triphenyl propan-2-yl phosphanium iodide, i-propyl triphenylphosphonium iodide, isopropyl triphenylphosphonium iodide, acmc-209gck, ksc493k7j, isopropyltriphenylphosphine iodide
PubChem CID 2723783
IUPAC Name triphenyl(propan-2-yl)phosphanium iodide
SMILES [I-].CC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 199°C
Quantity 5 g
Formula Weight 432.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropyltriphenylphosphonium Iodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.