Learn More
Enzo Life Sciences Isradipine (10mg). CAS: 75695-93-1
SDP
Supplier:  Enzo Life Sciences BMLCA2130010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Potent and selective L-type calcium channel blocker. A clinically useful antihypertensive agent which has anti-atherosclerotic effects and improves endothelium-mediated NO-dependent vasodilation. Displays neuroprotective activity in a rat model of focal ischemia. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml) or 100% ethanol (15 mg/ml). Long Term Storage: +4°C.
Catalog No. 50-201-0363
missing translation for 'QuantityLeft'