Learn More
TARGETMOL CHEMICALS INC ISRADIPINE 50MG
Supplier: TARGETMOL CHEMICALS INC T0957
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isradipine (PN 200-110) is a dihydropyridine calcium channel blockers with antihypertensive and vasodilator activities. Isradipine blocks the calcium entry through the calcium ion channels of coronary and peripheral vascular smooth muscle thereby dilating coronary arteries and peripheral arterioles. This increases oxygen delivery due to an increased blood flow and reduces oxygen requirements due to decreasing in total peripheral resistance. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.