Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

J 113863, Tocris Bioscience™

Catalog No. p-4784050 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent CCR1 chemokine receptor antagonist

Potent chemokine receptor 1 (CCR1) antagonist (IC50 values are 0.9 and 5.8 nM for human and mouse CCR1 receptors respectively). Also displays high selectivity for human but not mouse CCR3 receptors (IC50 values are 0.58 and 460 nM respectively). Improves paw inflammation, joint damage and dramatically reduces cell infiltration into joints in collagen-induced arthritis in mice.

Isomer also available.

Chemical Identifiers

CAS 353791-85-2
Molecular Formula C30H37Cl2IN2O2
Molecular Weight (g/mol) 655.442
InChI Key FOAFBMYSXIGAOX-UHFFFAOYSA-N
Synonym 1-cyclooct-1-en-1-yl methyl-4-2,7-dichloro-9h-xanthene-9-carbonyl amino-1-ethylpiperidin-1-ium iodide
PubChem CID 70296607
IUPAC Name 2,7-dichloro-N-[1-(cycloocten-1-ylmethyl)-1-ethylpiperidin-1-ium-4-yl]-9H-xanthene-9-carboxamide;iodide
SMILES CC[N+]1(CCC(CC1)NC(=O)C2C3=C(C=CC(=C3)Cl)OC4=C2C=C(C=C4)Cl)CC5=CCCCCCC5.[I-]

Specifications

Quantity 1 mg
Formula Weight 655.44
Percent Purity >98%
Chemical Name or Material J 113863
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.