Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Janelia Fluor™ 635, Tetrazine

Catalog No. 8134200U Shop All R&D Systems Products
Click to view available options
:
200 μg

Red dye (Reactive group: tetrazine). Application: confocal microscopy, super resolution microscopy including dSTORM & STED. Suitable for live cell imaging

Key Information: Janelia Fluor™ 635, Tetrazine is a red fluorogenic fluorescent dye; supplied with a tetrazine reactive handle for copper-free click chemistry. Suitable for live-cell imaging. Application: Suitable for flow cytometry, confocal microscopy, super resolution microscopy (SRM) techniques including dSTORM (in both live and fixed cells). Janelia Fluor™ 635, Tetrazine is cell permeable. Properties and Photophysical Data: Excitation and emission maxima (λ) are 645 nm and 663 nm, respectively. Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.

Specifications

Chemical Name or Material 4-((4-(1,2,4,5-Tetrazin-3-yl)benzyl)carbamoyl)-2-(3-(3-fluoroazetidin-1-ium-1-ylidene)-7-(3-fluoroazetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)benzoate
Quantity 200 μg
Target Fluorescent Dyes
Molecular Formula C38H33F2N7O3Si
Purity 0.9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.